3-Amino-6-methoxypyridazine
Supplier: Apollo Scientific
Warning
OR14965-1G OR14965-250MG
APOSOR14965-1GEA 68.3 EUR
APOSOR14965-1G APOSOR14965-250MG
3-Amino-6-methoxypyridazine
3-Amino-6-methoxypyridazine
Formula: C₅H₇N₃O
MW: 125.13 g/mol
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01646127
CAS Number: 7252-84-8
EINECS: 230-670-1