4,5-Dichloro-3(2H)-pyridazinone
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one
OR14973-100G OR14973-25G
APOSOR14973-100GEA 0 EUR
APOSOR14973-100G APOSOR14973-25G
4,5-Dichloro-3(2H)-pyridazinone
4,5-Dichloro-3(2H)-pyridazinone
Formula: C₄H₂Cl₂N₂O
MW: 164.98 g/mol
Melting Pt: 200…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00051504
CAS Number: 932-22-9