2,5-Dichloroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Dichloro-2-methoxybenzene
OR15004-25G OR15004-5G
APOSOR15004-25GEA 52.2 EUR
APOSOR15004-25G APOSOR15004-5G
2,5-Dichloroanisole
2,5-Dichloroanisole
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 128…132 °C/3.5mmHg
Melting Pt: 24
Density: 1.33 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061123
CAS Number: 1984-58-3
EINECS: 217-852-6