4-(Isopropylamino)diphenylamine
Supplier: Apollo Scientific
Warning
Synonyms: N1-Isopropyl-N4-phenylbenzene-1,4-diamine, N-Isopropyl-N'-phenyl-p-phenylenediamine
OR15007-100G OR15007-25G
APOSOR15007-100GEA 29 EUR
APOSOR15007-100G APOSOR15007-25G
4-(Isopropylamino)diphenylamine
4-(Isopropylamino)diphenylamine
Formula: C₁₅H₁₈N₂
MW: 226.32 g/mol
Boiling Pt: 161 °C/1mmHg
Melting Pt: 74…80 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00026348
CAS Number: 101-72-4
EINECS: 202-969-7
UN: 3077
ADR: 9+EHS III