2-Chloro-p-toluidine
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
OR15009-100G OR15009-25G OR15009-500G
APOSOR15009-100GEA 72.5 EUR
APOSOR15009-100G APOSOR15009-25G APOSOR15009-500G
2-Chloro-p-toluidine
2-Chloro-p-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 223…225 °C
Melting Pt: 7 °C
Density: 1.151 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007666
CAS Number: 615-65-6
EINECS: 210-440-7
UN: 3429
ADR: 6.1 III