2-Bromo-5-methylthiophene
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol
OR15029-100G OR15029-25G OR15029-5G
APOSOR15029-100GEA 128 EUR
APOSOR15029-100G APOSOR15029-25G APOSOR15029-5G
2-Bromo-5-methylthiophene
2-Bromo-5-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 65 °C/15mmHg
Melting Pt: 58
Density: 1.552 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130103
CAS Number: 765-58-2
EINECS: 212-151-1