2-Bromo-4-chloroaniline 98%
Supplier: Apollo Scientific
Warning
OR15032-10G OR15032-250G OR15032-50G
APOSOR15032-10GEA 21.9 EUR
APOSOR15032-10G APOSOR15032-250G APOSOR15032-50G
2-Bromo-4-chloroaniline 98%
2-Bromo-4-chloroaniline
Formula: C₆H₅BrClN
Boiling Pt: 127 °C/20mmHg
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00041313
CAS Number: 873-38-1
EINECS: 212-837-0