2-Amino-4-chlorobenzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-2-cyanoaniline
OR15043-1KG OR15043-250G OR15043-50G
APOSOR15043-1KGEA 560 EUR
APOSOR15043-1KG APOSOR15043-250G APOSOR15043-50G
2-Amino-4-chlorobenzonitrile
2-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00035927
CAS Number: 38487-86-4
EINECS: 253-967-8