tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 3-[(Dimethylamino)methylene]piperidin-4-one, N1-BOC protected
OR15082-10G OR15082-1G OR15082-5G
APOSOR15082-10GEA 219 EUR
APOSOR15082-10G APOSOR15082-1G APOSOR15082-5G
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD08443986
CAS Number: 157327-41-8