2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Chlorophenyl)-4-hydroxy-5-phenyl-1,3-thiazole
OR15157-1G OR15157-500MG
APOSOR15157-1GEA 40.6 EUR
APOSOR15157-1G APOSOR15157-500MG
2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol
2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol
Formula: C₁₅H₁₀ClNOS
MW: 287.77 g/mol
Melting Pt: 252…254 °C
Storage Temperature: Ambient
MDL Number: MFCD08444002
CAS Number: 65752-50-3