3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde
Supplier: Apollo Scientific
Warning
OR15166-10G OR15166-1G OR15166-5G
APOSOR15166-10GEA 1350 EUR
APOSOR15166-10G APOSOR15166-1G APOSOR15166-5G
3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde
3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde
Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD08443964
CAS Number: 924868-83-7