4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol
Supplier: Apollo Scientific
Synonyms:
2,6-Diisopropyl-4-[(dimethylamino)methyl]phenol
OR15178-10G
OR15178-1G
OR15178-5G
APOSOR15178-10GEA
894
EUR
APOSOR15178-10G
APOSOR15178-1G
APOSOR15178-5G
4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol
4-[(Dimethylamino)methyl]-2,6-diisopropylbenzenol
Formula:
C₁₅H₂₅NO MW: 235.37 g/mol Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00514869 CAS Number: 4918-95-0 |