Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Supplier: Apollo Scientific
Warning
OR15199-1G OR15199-5G
APOSOR15199-1GEA 55.1 EUR
APOSOR15199-1G APOSOR15199-5G
Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Ethyl-2-methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Formula: C₁₂H₁₂N₂O₆
MW: 280.24 g/mol
Melting Pt: 173…174 °C
Storage Temperature: Ambient
MDL Number: MFCD08443979
CAS Number: 154365-37-4