2-Methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylic acid
Supplier: Apollo Scientific
OR15231-1G OR15231-5G
APOSOR15231-1GEA 336 EUR
APOSOR15231-1G APOSOR15231-5G
2-Methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylic acid
2-Methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylic acid
Formula: C₉H₈N₂O₄
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD04124560
CAS Number: 154365-45-4