2-(4-Methylphenoxy)nicotinaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Methylphenoxy)pyridine-3-carboxaldehyde
OR15232-1G OR15232-5G
APOSOR15232-1GEA 163 EUR
APOSOR15232-1G APOSOR15232-5G
2-(4-Methylphenoxy)nicotinaldehyde
2-(4-Methylphenoxy)nicotinaldehyde
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00794346
CAS Number: 478030-89-6