2',4'-Dimethoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,4-Dimethoxyphenyl)ethan-1-one
OR1525-25G OR1525-1G OR1525-5G
APOSOR1525-25GEA 43.5 EUR
APOSOR1525-25G APOSOR1525-1G APOSOR1525-5G
2',4'-Dimethoxyacetophenone 98%
2',4'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 287…289 °C
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008727
CAS Number: 829-20-9
EINECS: 212-587-2