4-Chloro-7-azaindole
Supplier: Apollo Scientific
Danger
OR15256-5G
APOSOR15256-5GEA 24.8 EUR
APOSOR15256-5G
4-Chloro-7-azaindole
4-Chloro-7-azaindole
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 176…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272232
CAS Number: 55052-28-3
UN: 2811
ADR: 6.1,III