2-(tert-Butyl)-1H-indole 95+%
Supplier: Apollo Scientific
Warning
OR15275-5G
APOSOR15275-5GEA 329 EUR
APOSOR15275-5G
2-(tert-Butyl)-1H-indole 95+%
2-(tert-Butyl)-1H-indole
Formula: C₁₂H₁₅N
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD04004060
CAS Number: 1805-65-8