Ethyl 2-aminonicotinate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-aminonicotinate
OR15285-25G OR15285-5G
APOSOR15285-25GEA 72.5 EUR
APOSOR15285-25G APOSOR15285-5G
Ethyl 2-aminonicotinate 98%
Ethyl 2-aminonicotinate
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 266.75 °C
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01569452
CAS Number: 13362-26-0