2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid
Supplier: Apollo Scientific
Warning
OR15323-1G OR15323-250MG OR15323-500MG
APOSOR15323-1GEA 445 EUR
APOSOR15323-1G APOSOR15323-250MG APOSOR15323-500MG
2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid
2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid
Formula: C₁₆H₁₃ClO₃
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00664378
CAS Number: 344280-12-2