N-Boc-ethylenediamine 97+%
Supplier: Apollo Scientific
Danger
Synonyms: N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane
OR1533-25G OR1533-100G
APOSOR1533-25GEA 95.6 EUR
APOSOR1533-25G APOSOR1533-100G
N-Boc-ethylenediamine 97+%
N-Boc-ethylenediamine
Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 72…80 °C/0.1mmHg
Density: 1.012 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8 III