4-(4-Nitrophenyl)piperidine 98+%
Supplier: Apollo Scientific
|
Warning
|
OR15358-100MG
OR15358-250MG
APOSOR15358-100MGEA
21.9
EUR
APOSOR15358-100MG
APOSOR15358-250MG
4-(4-Nitrophenyl)piperidine 98+%
4-(4-Nitrophenyl)piperidine
Formula:
C₁₁H₁₄N₂O₂ Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD06804549 CAS Number: 26905-03-3 |