4-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonylchloride
Supplier: Apollo Scientific
Danger
OR15376-1G OR15376-5G
APOSOR15376-1GEA 243 EUR
APOSOR15376-1G APOSOR15376-5G
4-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonylchloride
4-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonylchloride
Formula: C₁₀H₉ClN₂O₂S
MW: 256.71 g/mol
Melting Pt: 89.5…91 °C
Storage Temperature: Ambient
MDL Number: MFCD09817463
CAS Number: 916766-81-9
UN: 3261
ADR: 8,III