3-Bromoadamantane-1-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR1538-1G OR1538-5G
APOSOR1538-1GEA 21.9 EUR
APOSOR1538-1G APOSOR1538-5G
3-Bromoadamantane-1-carboxylic acid 97%
3-Bromoadamantane-1-carboxylic acid
Formula: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167820
CAS Number: 21816-08-0