2-Piperidinonicotinaldehyde 95%
Supplier: Apollo Scientific
Warning
OR15429-1G OR15429-500MG
APOSOR15429-1GEA 538 EUR
APOSOR15429-1G APOSOR15429-500MG
2-Piperidinonicotinaldehyde 95%
2-Piperidinonicotinaldehyde
Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865118
CAS Number: 34595-22-7