4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide
Supplier: Apollo Scientific
Synonyms: 4,5-Dibromo-2-[(3-hydroxyprop-1-yl)carbamoyl]-1H-pyrrole
OR15433-10G OR15433-1G OR15433-5G
APOSOR15433-10GEA 1690 EUR
APOSOR15433-10G APOSOR15433-1G APOSOR15433-5G
4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide
4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide
Formula: C₈H₁₀Br₂N₂O₂
MW: 326 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD06200910
CAS Number: 186956-98-9