2-(4-Chlorophenylthio)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 2-[(4-Chlorophenyl)sulphanyl]-1-ethanol, 4-Chlorophenyl-2-hydroxyethyl sulphide
OR15484-1G OR15484-250MG OR15484-5G
APOSOR15484-1GEA 109 EUR
APOSOR15484-1G APOSOR15484-250MG APOSOR15484-5G
2-(4-Chlorophenylthio)ethanol
2-(4-Chlorophenylthio)ethanol
Formula: C₈H₉ClOS
MW: 188.68 g/mol
Boiling Pt: 305
Melting Pt: 41
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00014037
CAS Number: 13457-98-2