3-Chloro-o-cresol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Chloro-2-methylphenol, 1-Chloro-3-hydroxy-2-methylbenzene
OR1558-100G OR1558-25G OR1558-5G
APOSOR1558-100GEA 770 EUR
APOSOR1558-100G APOSOR1558-25G APOSOR1558-5G
3-Chloro-o-cresol 98%
3-Chloro-o-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 225 °C
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00079738
CAS Number: 3260-87-5
EINECS: 221-861-0