2-Methoxy-6-(tributylstannyl)pyridine
Supplier: Apollo Scientific
Danger
OR15611-1G OR15611-500MG
APOSOR15611-1GEA 105 EUR
APOSOR15611-1G APOSOR15611-500MG
2-Methoxy-6-(tributylstannyl)pyridine
2-Methoxy-6-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
Storage Temperature: Refrigerator
MDL Number: MFCD09025803
CAS Number: 164014-94-2
UN: 2788
ADR: 6.1 III