(S)-(-)-1-Boc-3-methylpiperazine
Supplier: Apollo Scientific
Danger
Synonyms: (S)-tert-Butyl 3-methylpiperazine-1-carboxylate, (3S)-3-Methylpiperazine, N1-BOC protected
OR15643-25G
APOSOR15643-25GEA 24.8 EUR
APOSOR15643-25G
(S)-(-)-1-Boc-3-methylpiperazine
(S)-(-)-1-Boc-3-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 38…45 °C
Storage Temperature: Ambient
MDL Number: MFCD02683204
CAS Number: 147081-29-6