(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-1-BOC-3-(hydroxymethyl)piperidine, (R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(R)-N-BOC-PIPERIDINE METHANOL, 1-Piperidinecarboxylicacid, 3-(hydroxymethyl)-, 1,1-dimethylethyl ester, (3R)-
OR15673-1G OR15673-25G OR15673-5G
APOSOR15673-1GEA 21.9 EUR
APOSOR15673-1G APOSOR15673-25G APOSOR15673-5G
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate
Formula: C₁₁H₂₁NO₃
MW: 215.293 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD02683202
CAS Number: 140695-85-8