1-Boc-3-(methylamino)piperidine
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine
OR15675-25G OR15675-5G OR15675-100G
APOSOR15675-25GEA 255 EUR
APOSOR15675-25G APOSOR15675-5G APOSOR15675-100G
1-Boc-3-(methylamino)piperidine
1-Boc-3-(methylamino)piperidine
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 95…99 °C/ 0.6mmHg
Density: 0.983 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114980
CAS Number: 392331-89-4
UN: 2810
ADR: 6.1 III