3-Boc-aminopyrrolidine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine
OR15678-5G OR15678-1G OR15678-25G
APOSOR15678-5GEA 72.5 EUR
APOSOR15678-5G APOSOR15678-1G APOSOR15678-25G
3-Boc-aminopyrrolidine 95%
3-Boc-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Melting Pt: 63…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059040
CAS Number: 99724-19-3