4-Amino-2,6-dimethylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dimethylpyridin-4-amine
OR15746-1G OR15746-5G
APOSOR15746-1GEA 46.4 EUR
APOSOR15746-1G APOSOR15746-5G
4-Amino-2,6-dimethylpyridine
4-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Melting Pt: 186…187 °C
Density: 1.039 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130078
CAS Number: 3512-80-9
EINECS: 225-515-1