5-Aminobenzotriazole 98%
Supplier: Apollo Scientific
Warning
OR15753-10G OR15753-5G
APOSOR15753-10GEA 190 EUR
APOSOR15753-10G APOSOR15753-5G
5-Aminobenzotriazole 98%
5-Aminobenzotriazole
Formula: C₆H₆N₄
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00047209
CAS Number: 3325-11-9