3-Amino-2,6-dimethoxypyridine 95+%
Supplier: Apollo Scientific
Warning
OR1585-1G OR1585-250MG OR1585-5G
APOSOR1585-1GEA 65.4 EUR
APOSOR1585-1G APOSOR1585-250MG APOSOR1585-5G
3-Amino-2,6-dimethoxypyridine 95+%
3-Amino-2,6-dimethoxypyridine
Formula: C7H10N2O2
MW: 154.1665 g/mol
Boiling Pt: 116…117 °C/10mmHg
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00832865
CAS Number: 28020-37-3