2-Chlorothiophenol 99%
Supplier: Apollo Scientific
Danger
Synonyms: o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol
OR1590-100G OR1590-25G OR1590-5G
APOSOR1590-100GEA 95.7 EUR
APOSOR1590-100G APOSOR1590-25G APOSOR1590-5G
2-Chlorothiophenol 99%
2-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 204…206 °C
Density: 1.275 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004830
CAS Number: 6320-03-2
EINECS: 228-667-5
UN: 3265
ADR: 8,II