(2S,3S)-(+)-2,3-Butanediol
Supplier: Apollo Scientific
Warning
Synonyms: (2S,3S)-Butane-2,3-diol, (S,S)-(+)-2,3-Butanediol, (2S,3S)-(+)-2,3-Butylene glykol, (S,S)-(+)-2,3-Dihydroxy butane, (2S,3S)-(+)-2,3-Dihydroxy butane, (S,S)-(+)-2,3-Butylene glykol, L(+)-2,3-Butanediol
OR15919-1G OR15919-5G OR15919-250MG
APOSOR15919-1GEA 37.7 EUR
APOSOR15919-1G APOSOR15919-5G APOSOR15919-250MG
(2S,3S)-(+)-2,3-Butanediol
(2S,3S)-(+)-2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 179…182 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00063648
CAS Number: 19132-06-0