(1R,2R)-(+)-1,2-Diphenylethylenediamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1R,2R)-(+)-1,2-Diphenylethylenediamine, (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, (1R,2R)-1,2-Diphenylethane-1,2-diamine, (R,R)-DPEN, (+)-((R,R)-1,2-Diamino-2-phenylethyl)benzene
OR15922-5G OR15922-25G OR15922-100G
APOSOR15922-5GEA 21.9 EUR
APOSOR15922-5G APOSOR15922-25G APOSOR15922-100G
(1R,2R)-(+)-1,2-Diphenylethylenediamine 98%
(1R,2R)-(+)-1,2-Diphenylethylenediamine
Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
Melting Pt: 79…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082769
CAS Number: 35132-20-8