4,5,6-Trichloropyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR15924-1G
OR15924-10G
APOSOR15924-1GEA
52.2
EUR
APOSOR15924-1G
APOSOR15924-10G
4,5,6-Trichloropyrimidine
4,5,6-Trichloropyrimidine
Formula:
C₄HCl₃N₂ MW: 183.4231 g/mol Boiling Pt: 87 °C/12mmHg Melting Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458457 CAS Number: 1780-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...