tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate, N-Boc-2,5-dihydropyrrole, N-BOC-3-Pyrroline
OR15933-100G OR15933-500G
APOSOR15933-100GEA 97.3 EUR
APOSOR15933-100G APOSOR15933-500G
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Formula: C₉H₁₅NO₂
MW: 169.224 g/mol
Boiling Pt: 208 °C
Density: 0.981 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD01863512
CAS Number: 73286-70-1