Methyl-2-bromofuran-3-carboxylate 97%
Supplier: Apollo Scientific
Warning
OR15964-1G OR15964-250MG OR15964-100MG
APOSOR15964-1GEA 273 EUR
APOSOR15964-1G APOSOR15964-250MG APOSOR15964-100MG
Methyl-2-bromofuran-3-carboxylate 97%
Methyl-2-bromofuran-3-carboxylate
Formula: C₆H₅BrO₃
MW: 205.01 g/mol
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD09879944
CAS Number: 197846-06-3