5-Bromo-2-chlorotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1-chloro-2-methylbenzene, 1-Bromo-4-chloro-3-methylbenzene
OR1599-25G OR1599-100G OR1599-5G
APOSOR1599-25GEA 41.4 EUR
APOSOR1599-25G APOSOR1599-100G APOSOR1599-5G
5-Bromo-2-chlorotoluene
5-Bromo-2-chlorotoluene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 90…93 °C/10mmHg
Density: 1.549 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00018503
CAS Number: 54932-72-8