1,10-Phenanthroline-5,6-dione 98%
Supplier: Apollo Scientific
Warning
OR16008-1G OR16008-25G OR16008-5G
APOSOR16008-1GEA 58 EUR
APOSOR16008-1G APOSOR16008-25G APOSOR16008-5G
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione
Formula: C₁₂H₆N₂O₂
MW: 210.19 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014473
CAS Number: 27318-90-7