4-Chlorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: p-Chlorobenzoyl chloride
OR16400-100G OR16400-25G
APOSOR16400-100GEA 33.5 EUR
APOSOR16400-100G APOSOR16400-25G
4-Chlorobenzoyl chloride 98%
4-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 41944 °C
Density: 1.377 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS Number: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II