Ethyl-3-(dimethylamino)acrylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-(dimethylamino)acrylate, 3-(Dimethylamino)acrylic Acid Ethyl Ester, Ethyl 3-(N,N-dimethylamino)acrylate
OR16403-100G OR16403-25G
APOSOR16403-100GEA 58 EUR
APOSOR16403-100G APOSOR16403-25G
Ethyl-3-(dimethylamino)acrylate
Ethyl-3-(dimethylamino)acrylate
Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 118…121 °C/10mmHg
Melting Pt: 17…18 °C
Density: 1 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00144269
CAS Number: 924-99-2