N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Diethylamino)phenylboronic Acid Pinacol Ester, 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR16404-1G OR16404-250MG OR16404-5G
APOSOR16404-1GEA 219 EUR
APOSOR16404-1G APOSOR16404-250MG APOSOR16404-5G
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formula: C₁₆H₂₆BNO₂
MW: 275.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000956
CAS Number: 920304-57-0