2,2,2-Trichloroethyl chloroformate 95%
Supplier: Apollo Scientific
Danger
OR16412-100G OR16412-25G OR16412-500G
APOSOR16412-100GEA 116 EUR
APOSOR16412-100G APOSOR16412-25G APOSOR16412-500G
2,2,2-Trichloroethyl chloroformate 95%
2,2,2-Trichloroethyl chloroformate
Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 171…172 °C
Melting Pt: 0…2 °C
Density: 1.539 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1+8 II