1-Chloroethyl chloroformate 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloroethyl carbonochloridate
OR16433-100G OR16433-25G OR16433-500G
APOSOR16433-100GEA 65.4 EUR
APOSOR16433-100G APOSOR16433-25G APOSOR16433-500G
1-Chloroethyl chloroformate 98%
1-Chloroethyl chloroformate
Formula: C₃H₄Cl₂O₂
Boiling Pt: 118…121 °C
Melting Pt: –65 °C
Density: 1.325 g/cm³ (25 °C)
Flash Pt: 40.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1+3+8,II