1,1'-Bis(di-tert-butylphosphino)ferrocene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF
OR16471-5G OR16471-10G OR16471-1G
APOSOR16471-5GEA 26.1 EUR
APOSOR16471-5G APOSOR16471-10G APOSOR16471-1G
1,1'-Bis(di-tert-butylphosphino)ferrocene 98%
1,1'-Bis(di-tert-butylphosphino)ferrocene
Formula: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD01630818
CAS Number: 84680-95-5